Assay Details
Target Gene Details
Entrez Gene ID: | 73341 |
Gene Name: | Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6 |
Gene Aliases: |
1600028C08Rik, 1700038J06Rik, 4930592P22Rik, alpha-PIX |
Location: |
Chr.X:57231485-57338756 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef6 | NM_152801.2 | 12 | 1552 | NP_690014.2 |
| AF393831.1 | 12 | 1552 | AAM94903.1 | |
| AK006635.1 | 7 | 699 | ||
| AK040468.1 | 12 | 1374 | BAC30599.1 |
More Information
Set Membership: |
|

Genomic Map