Assay Details
Target Gene Details
Entrez Gene ID: | 108062 |
Gene Name: | cleavage stimulation factor, 3' pre-RNA subunit 2 |
Gene Aliases: |
C630034J23Rik, Cstf64, mir-3112 |
Location: |
Chr.X:134059139-134086822 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cstf2 | NM_001290398.1 | 4 | 558 | NP_001277327.1 |
| NM_001290399.1 | 4 | 558 | NP_001277328.1 | |
| NM_133196.6 | 4 | 558 | NP_573459.1 | |
| AF317552.1 | 4 | 526 | AAG31814.1 | |
| AK032817.1 | 4 | 540 | BAC28037.1 | |
| AK049971.1 | 4 | 548 | ||
| AK088260.1 | 4 | 386 | BAC40243.1 | |
| AK147438.1 | 3 | 324 | ||
| BC036719.1 | 4 | 530 | AAH36719.1 | |
| EU616680.1 | 4 | 526 | ||
| EU616681.1 | 4 | 526 | ||
| EU616682.1 | 4 | 526 |
More Information
Set Membership: |
|

Genomic Map