Assay Details
Target Gene Details
Entrez Gene ID: | 271564 |
Gene Name: | vacuolar protein sorting 13A (yeast) |
Gene Aliases: |
4930425F11, 4930516E05Rik, 4930543C13Rik, 9930023P20, Chac, Chorein, D330038K10Rik, mKIAA0986 |
Location: |
Chr.19:16612194-16781139 on Build GRCm38 |
Assay Gene Location: | Within Exon 56 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps13a | NM_173028.4 | 56 | 8023 | NP_766616.2 |
| AB115421.1 | 56 | 8018 | BAD89296.1 | |
| AK029649.1 | 10 | 1778 | BAC26546.1 | |
| AK030073.1 | 9 | 1247 | ||
| AK084751.1 | 2 | 218 | ||
| AK135983.1 | 13 | 2177 | BAE22761.1 | |
| AK220362.1 | 9 | 1440 | BAD90423.1 | |
| BC050055.1 | 3 | 433 | AAH50055.1 | |
| BC062130.1 | 12 | 1921 | AAH62130.1 |
More Information
Set Membership: |
|

Genomic Map