Assay Details
Target Gene Details
Entrez Gene ID: | 75013 |
Gene Name: | RIKEN cDNA 4930502E18 gene |
Gene Aliases: |
Ct55 |
Location: |
Chr.X:53724831-53738565 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930502E18Rik | NM_029142.1 | NP_083418.1 | ||
| AK015678.1 | BAB29927.1 | |||
| BC115663.1 | ||||
| BC115664.1 |

Genomic Map