Assay Details
Target Gene Details
Entrez Gene ID: | 12705 |
Gene Name: | Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1 |
Gene Aliases: |
AI316840, AU019144, Msg1 |
Location: |
Chr.X:102247378-102252181 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cited1 | NM_001276466.1 | NP_001263395.1 | ||
| NM_001276473.1 | NP_001263402.1 | |||
| NM_001276474.1 | NP_001263403.1 | |||
| NM_007709.4 | NP_031735.1 | |||
| AK148042.1 | BAE28308.1 | |||
| BC052030.1 | AAH52030.1 | |||
| CX237826.1 | ||||
| U65091.1 | AAC53048.1 |
More Information
Set Membership: |
|

Genomic Map