Assay Details
Target Gene Details
Entrez Gene ID: | 54004 |
Gene Name: | diaphanous homolog 2 (Drosophila) |
Gene Aliases: |
Dia3, Diaph2, Drf2, E430022I22Rik |
Location: |
Chr.X:129749668-130465834 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 22 - Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Diap2 | NM_172493.2 | NP_766081.1 | ||
| AB713187.1 | ||||
| AK083254.1 | BAC38832.1 | |||
| AK088648.1 | BAC40476.1 | |||
| AK153986.1 | BAE32296.1 | |||
| AY312280.1 | AAQ82539.1 | |||
| BC100759.1 | AAI00760.1 | |||
| Y15910.1 | CAA75871.1 |

Genomic Map