Assay Details
Target Gene Details
Entrez Gene ID: | 54652 |
Gene Name: | calcium channel, voltage-dependent, alpha 1F subunit |
Gene Aliases: |
A930034B14, Cav1.4, Sfc17, nerg1, nob2 |
Location: |
Chr.X:7606982-7635196 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna1f | NM_019582.2 | 13 | 1689 | NP_062528.2 |
| AF192497.1 | 13 | 1694 | AAF86764.1 | |
| AJ579852.2 | 13 | 1646 | CAE30292.1 | |
| AK044695.1 | 13 | 1673 | BAC32037.1 |
More Information
Set Membership: |
|

Genomic Map