Assay Details
Target Gene Details
Entrez Gene ID: | 245474 |
Gene Name: | dyskeratosis congenita 1, dyskerin |
Gene Aliases: |
BC068171 |
Location: |
Chr.X:75095854-75109777 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dkc1 | NM_001030307.2 | NP_001025478.1 | ||
| AK136418.1 | BAE22970.1 | |||
| AK144139.1 | ||||
| AK166105.1 | BAE38575.1 | |||
| AK167973.1 | BAE39967.1 | |||
| BC099966.1 | AAH99966.1 |
More Information
Set Membership: |
|

Genomic Map