Assay Details
Target Gene Details
Entrez Gene ID: | 333564 |
Gene Name: | fibronectin type III domain containing 3C1 |
Gene Aliases: |
Fndc3c, Gm784 |
Location: |
Chr.X:106420041-106485576 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fndc3c1 | NM_001007580.1 | NP_001007581.1 | ||
| BC076625.1 | AAH76625.1 | |||
| BC085176.1 | AAH85176.1 | |||
| DQ192038.1 | ABA82151.1 |
More Information
Set Membership: |
|

Genomic Map