Assay Details
Target Gene Details
Entrez Gene ID: | 11704 |
Gene Name: | amelogenin, X-linked |
Gene Aliases: |
ALGN, AMGL, AMGX, Amel, Amg, LRAP, M100888, Rgsc888 |
Location: |
Chr.X:169175044-169187287 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amelx | NM_001081978.2 | NP_001075447.1 | ||
| NM_001290371.1 | NP_001277300.1 | |||
| NM_009666.4 | NP_033796.1 | |||
| AK029358.1 | BAC26415.1 | |||
| BC059090.1 | AAH59090.1 | |||
| CD773419.1 | ||||
| D31768.1 | BAA06546.1 | |||
| D31769.1 | BAA06547.1 | |||
| KC731591.1 | ||||
| S74899.1 | AAB33093.1 |
Target Gene Details
Entrez Gene ID: | 11856 |
Gene Name: | Rho GTPase activating protein 6 |
Gene Aliases: |
AI504284, RhoGAPX-1 |
Location: |
Chr.X:168795094-169304440 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap6 | NM_001287530.1 | NP_001274459.1 | ||
| NM_009707.4 | NP_033837.2 | |||
| NM_178754.3 | NP_848869.1 | |||
| NR_109847.1 | ||||
| AK047827.1 | BAC33167.1 | |||
| AK048162.1 | BAC33263.1 | |||
| AK048507.1 | BAC33352.1 | |||
| AK077996.1 | BAC37093.1 | |||
| AK079651.1 | ||||
| AK164501.1 | BAE37813.1 | |||
| AK166015.1 | BAE38520.1 | |||
| AK170575.1 | ||||
| AK171337.1 | ||||
| AK171677.1 | BAE42606.1 |
More Information
Set Membership: |
|

Genomic Map