Assay Details
Target Gene Details
Entrez Gene ID: | 170735 |
Gene Name: | arrestin 3, retinal |
Gene Aliases: |
Arr4, Car, Carfl, Carr |
Location: |
Chr.X:100605480-100618953 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arr3 | NM_133205.3 | NP_573468.1 |
More Information
Set Membership: |
|

Genomic Map