Assay Details
Target Gene Details
Entrez Gene ID: | 20371 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
JM2, scurfin, sf |
Location: |
Chr.X:7579676-7595243 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxp3 | NM_001199347.1 | 4 | 397 | NP_001186276.1 |
| NM_001199348.1 | 3 | 255 | NP_001186277.1 | |
| NM_054039.2 | 3 | 330 | NP_473380.1 | |
| AF277991.1 | 3 | 255 | AAG53605.1 | |
| AF277992.1 | 3 | 313 | AAG53606.1 | |
| AK157447.1 | 4 | 398 | BAE34089.1 | |
| AY357712.1 | 2 | 113 | AAR11305.1 | |
| AY357713.1 | 2 | 113 | AAR11306.1 | |
| BC132333.1 | 2 | 101 | ||
| BC132335.1 | 2 | 101 | ||
| DQ387959.1 | 4 | 397 | ||
| KF471319.1 | 2 | 97 |

Genomic Map