Assay Details
Target Gene Details
Entrez Gene ID: | 15929 |
Gene Name: | isocitrate dehydrogenase 3 (NAD+), gamma |
Gene Aliases: |
- |
Location: |
Chr.X:73778963-73786953 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Idh3g | NM_008323.1 | NP_032349.1 | ||
| AJ748655.1 | CAG38689.1 | |||
| AK156309.1 | BAE33665.1 | |||
| AK166925.1 | BAE39120.1 | |||
| AK167131.1 | ||||
| AK168526.1 | BAE40405.1 | |||
| BC085179.1 | ||||
| BC119170.1 | ||||
| BC119172.1 | ||||
| U68564.1 | AAC53340.1 |
More Information
Set Membership: |
|

Genomic Map