Assay Details
Target Gene Details
Entrez Gene ID: | 77286 |
Gene Name: | NF-kappaB repressing factor |
Gene Aliases: |
9430034D17Rik, AW124770, C80287 |
Location: |
Chr.X:36886440-36903110 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkrf | NM_029891.2 | NP_084167.2 | ||
| AK020455.1 | BAB32107.1 | |||
| AK042690.1 | BAC31334.1 | |||
| BC138828.1 | ||||
| BC138829.1 |
More Information
Set Membership: |
|

Genomic Map