Assay Details
Target Gene Details
Entrez Gene ID: | 74131 |
Gene Name: | SAM and SH3 domain containing 3 |
Gene Aliases: |
1200013B08Rik, AW413946, SLY1 |
Location: |
Chr.X:48146451-48161565 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sash3 | NM_028773.3 | NP_083049.1 | ||
| AJ306422.1 | CAC67499.1 | |||
| AK004734.2 | BAB23516.1 | |||
| AK156202.1 | BAE33623.1 | |||
| AK171017.1 | BAE42187.1 | |||
| BC028773.1 | AAH28773.1 |

Genomic Map