Assay Details
Target Gene Details
Entrez Gene ID: | 18824 |
Gene Name: | proteolipid protein 2 |
Gene Aliases: |
mIMA4 |
Location: |
Chr.X:7668116-7671278 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plp2 | NM_019755.4 | NP_062729.1 | ||
| AB031292.1 | BAA83500.1 | |||
| AK003522.1 | BAB22835.1 | |||
| AK011282.1 | BAB27515.1 | |||
| AK012816.1 | ||||
| AK154228.1 | BAE32448.1 | |||
| BC083078.1 | AAH83078.1 | |||
| BC092095.1 | AAH92095.1 | |||
| BC119088.1 | ||||
| BC119090.1 |
More Information
Set Membership: |
|

Genomic Map