Assay Details
Target Gene Details
Entrez Gene ID: | 53623 |
Gene Name: | glutamate receptor, ionotropic, AMPA3 (alpha 3) |
Gene Aliases: |
2900064I19Rik, GluA3, GluR-C, GluR-K3, Glur-3, Glur3, Gluralpha3 |
Location: |
Chr.X:41401119-41678601 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gria3 | NM_001281929.1 | 15 | 2811 | NP_001268858.1 |
| NM_001290451.1 | 15 | 2583 | NP_001277380.1 | |
| NM_016886.4 | 15 | 2811 | NP_058582.3 | |
| NR_104053.1 | 16 | 2926 | ||
| AB022342.1 | 15 | 2601 | BAA37124.1 | |
| AB079072.1 | 15 | 2547 | BAE06153.1 | |
| AB102775.1 | 15 | 2547 | ||
| AB102776.1 | 14 | 2432 | ||
| AF483494.1 | 15 | 2547 | AAL90768.1 | |
| AF483495.1 | 15 | 2547 | AAL90769.1 | |
| AK135043.1 | 15 | 2565 | BAE22396.1 | |
| AK138615.1 | 14 | 3272 | ||
| AK220530.1 | 15 | 2598 | BAD90527.1 | |
| BC086678.1 | 15 | 2756 | ||
| BC129855.1 | 15 | 2601 | ||
| BC129856.1 | 15 | 2601 |
More Information
Set Membership: |
|

Genomic Map