Assay Details
Target Gene Details
Entrez Gene ID: | 54630 |
Gene Name: | prickle homolog 3 (Drosophila) |
Gene Aliases: |
Lmo6, Sfc16 |
Location: |
Chr.X:7657379-7668186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prickle3 | NM_001290624.1 | NP_001277553.1 | ||
| AK037217.1 | BAC29758.1 | |||
| AK083704.1 | BAC38998.1 | |||
| BC023671.1 | AAH23671.1 | |||
| BC049258.1 | AAH49258.1 |
More Information
Set Membership: |
|

Genomic Map