Assay Details
Target Gene Details
Entrez Gene ID: | 56526 |
Gene Name: | septin 6 |
Gene Aliases: |
2810035H17Rik, C920001C06Rik, Sep6, mKIAA0128 |
Location: |
Chr.X:36911272-36989695 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept6 | NM_001177323.2 | 5 | 855 | NP_001170794.1 |
| NM_001177324.1 | 5 | 858 | NP_001170795.1 | |
| NM_001253706.1 | 5 | 858 | NP_001240635.1 | |
| NM_019942.5 | 5 | 858 | NP_064326.2 | |
| AB023622.1 | 5 | 622 | BAA82838.1 | |
| AK012858.1 | 6 | 652 | BAB28516.1 | |
| AK048455.1 | 5 | 680 | BAC33342.1 | |
| AK088406.1 | 5 | 638 | BAC40335.1 | |
| AK088614.1 | 5 | 621 | BAC40453.1 | |
| AK160884.1 | 5 | 652 | BAE36069.1 | |
| AK162755.1 | 5 | 658 | BAE37050.1 | |
| AK172894.1 | 5 | 639 | BAD32172.1 | |
| BC010489.1 | 5 | 644 | AAH10489.1 |
More Information
Set Membership: |
|

Genomic Map