Assay Details
Target Gene Details
Entrez Gene ID: | 338351 |
Gene Name: | A kinase (PRKA) anchor protein 17B |
Gene Aliases: |
AA407619, AKAP-17B, B230333C21Rik, C230056F04Rik, PRKA17B, Sfrs17b, Srsf17b, Talia |
Location: |
Chr.X:36608173-36645471 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akap17b | NM_001081956.1 | NP_001075425.1 | ||
| AK046007.1 | BAC32570.2 | |||
| AK054091.1 | BAC35651.1 | |||
| AK136221.1 | ||||
| DV649862.1 |

Genomic Map