Assay Details
Target Gene Details
Entrez Gene ID: | 434768 |
Gene Name: | reproductive homeobox 8 |
Gene Aliases: |
Tox |
Location: |
Chr.X:37874776-37890766 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhox8 | NM_001004193.2 | NP_001004193.1 | ||
| AY329370.1 | AAQ94736.1 | |||
| BC139098.1 | ||||
| DQ058645.1 | AAY58256.1 |

Genomic Map