Assay Details
Target Gene Details
Entrez Gene ID: | 20400 |
Gene Name: | SH2 domain containing 1A |
Gene Aliases: |
Gm686, SAP |
Location: |
Chr.X:42502608-42522075 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh2d1a | NM_011364.3 | NP_035494.1 | ||
| AF072903.1 | AAC62629.1 | |||
| AF097632.1 | AAC95998.1 | |||
| AF097633.1 | AAC95999.1 | |||
| AK040076.1 | BAC30506.1 | |||
| BC065390.1 | AAH65390.1 | |||
| BC147367.1 | ||||
| BC147368.1 | ||||
| BC147807.1 | ||||
| BC147811.1 | ||||
| KF811030.1 |

Genomic Map