Assay Details
Target Gene Details
Entrez Gene ID: | 236792 |
Gene Name: | membrane magnesium transporter 1 |
Gene Aliases: |
9630048L06Rik, BC032271, Emc5, Mir5116, Tmem32, mir-5116 |
Location: |
Chr.X:56585512-56597919 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmgt1 | NM_146234.3 | NP_666346.1 | ||
| AK036253.1 | BAC29362.1 | |||
| AK043524.1 | BAC31567.1 | |||
| AK049689.1 | BAC33874.1 | |||
| AK163269.1 | ||||
| BC032271.1 | AAH32271.1 | |||
| EU069461.1 |
More Information
Set Membership: |
|

Genomic Map