Assay Details
Target Gene Details
Entrez Gene ID: | 20843 |
Gene Name: | stromal antigen 2 |
Gene Aliases: |
9230105L23Rik, B230112I07Rik, SA-2, SAP2 |
Location: |
Chr.X:42149322-42277186 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 21 - Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stag2 | NM_001077712.2 | NP_001071180.1 | ||
| NM_001290713.1 | NP_001277642.1 | |||
| NM_021465.4 | NP_067440.3 | |||
| AJ002636.1 | CAA05638.1 |
More Information
Set Membership: |
|

Genomic Map