Assay Details
Target Gene Details
Entrez Gene ID: | 93843 |
Gene Name: | pregnancy upregulated non-ubiquitously expressed CaM kinase |
Gene Aliases: |
Bstk3, CaMKIbeta2, CaMKlb2, Camk1b, Punc, caMKlb1 |
Location: |
Chr.X:73655992-73660127 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnck | NM_001199351.1 | NP_001186280.1 | ||
| NM_001199352.1 | NP_001186281.1 | |||
| NM_012040.3 | NP_036170.1 | |||
| AF181984.1 | AAF29157.1 | |||
| AK158755.1 | BAE34642.1 | |||
| BC051996.1 | AAH51996.1 | |||
| BC055891.1 | AAH55891.1 | |||
| CX243772.1 |
More Information
Set Membership: |
|

Genomic Map