Assay Details
Target Gene Details
Entrez Gene ID: | 20843 |
Gene Name: | stromal antigen 2 |
Gene Aliases: |
9230105L23Rik, B230112I07Rik, SA-2, SAP2 |
Location: |
Chr.X:42149322-42277186 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stag2 | NM_001077712.2 | 7 | 887 | NP_001071180.1 |
| NM_001290713.1 | 6 | 613 | NP_001277642.1 | |
| NM_021465.4 | 7 | 747 | NP_067440.3 | |
| AJ002636.1 | 6 | 589 | CAA05638.1 | |
| AK146718.1 | 7 | 747 | BAE27381.1 | |
| AK168899.1 | 7 | 747 | BAE40715.1 | |
| CO434048.1 | 5 | 434 |
More Information
Set Membership: |
|

Genomic Map