Assay Details
Target Gene Details
Entrez Gene ID: | 14933 |
Gene Name: | glycerol kinase |
Gene Aliases: |
D930012N15Rik, GK |
Location: |
Chr.X:85701937-85776819 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gyk | NM_001294138.1 | NP_001281067.1 | ||
| NM_001294140.1 | NP_001281069.1 | |||
| NM_008194.3 | NP_032220.1 | |||
| NM_212444.2 | NP_997609.1 | |||
| AK008186.1 | BAB25518.1 | |||
| AK044308.1 | BAC31861.1 | |||
| AK088373.1 | BAC40312.1 | |||
| AK169424.1 | BAE41166.1 | |||
| AK169525.1 | BAE41209.1 | |||
| BC003767.1 | AAH03767.1 | |||
| U48403.1 | AAC52824.1 |
More Information
Set Membership: |
|

Genomic Map