Assay Details
Target Gene Details
Entrez Gene ID: | 208431 |
Gene Name: | shroom family member 4 |
Gene Aliases: |
D430043L16Rik, Gm724, Shrm4 |
Location: |
Chr.X:6399938-6637452 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shroom4 | NM_001040459.1 | NP_001035549.1 | ||
| AK052531.1 | ||||
| AK173129.1 | BAD32407.1 | |||
| BC151022.1 | ||||
| DQ435686.1 | ABD98016.1 | |||
| DQ435687.1 | ABD98017.1 |
More Information
Set Membership: |
|

Genomic Map