Assay Details
Target Gene Details
Entrez Gene ID: | 209003 |
Gene Name: | RNA binding motif protein, X-linked 2 |
Gene Aliases: |
2700089A17Rik, 2810411A03Rik, CGI-79 |
Location: |
Chr.X:48695004-48710719 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbmx2 | NM_173376.3 | NP_775552.1 | ||
| AK076115.1 | BAC36195.1 | |||
| AK160232.1 | BAE35704.1 | |||
| AK168018.1 | BAE40004.1 | |||
| BC026976.1 | AAH26976.1 |

Genomic Map