Assay Details
Target Gene Details
Entrez Gene ID: | 19889 |
Gene Name: | retinitis pigmentosa 2 homolog (human) |
Gene Aliases: |
AI662636, Rp2 |
Location: |
Chr.X:20364481-20405653 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rp2h | NM_001290643.1 | NP_001277572.1 | ||
| NM_001290644.1 | NP_001277573.1 | |||
| NM_133669.5 | NP_598430.1 | |||
| NR_110966.1 | ||||
| NR_110967.1 | ||||
| AJ303371.1 | CAC21499.1 | |||
| AK017422.1 | ||||
| AK033954.1 | BAC28525.1 | |||
| AK044002.1 | BAC31734.1 | |||
| AK050418.1 | BAC34245.1 | |||
| AK076907.1 | BAC36524.1 | |||
| AK132713.1 | BAE21316.1 | |||
| AK162309.1 | BAE36846.1 | |||
| BC049698.1 | AAH49698.1 |
More Information
Set Membership: |
|

Genomic Map