Assay Details
Target Gene Details
Entrez Gene ID: | 15159 |
Gene Name: | holocytochrome c synthetase |
Gene Aliases: |
- |
Location: |
Chr.X:169311530-169320343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hccs | NM_008222.4 | NP_032248.3 | ||
| AK077566.1 | BAC36866.1 | |||
| AK143150.1 | ||||
| BC061466.1 | AAH61466.1 | |||
| BC062905.1 | AAH62905.1 | |||
| U36788.1 | AAB19008.1 |
More Information
Set Membership: |
|

Genomic Map