Assay Details
Target Gene Details
Entrez Gene ID: | 20964 |
Gene Name: | synapsin I |
Gene Aliases: |
Syn-1, Syn1-S |
Location: |
Chr.X:20860511-20920918 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syn1 | NM_001110780.1 | NP_001104250.1 | ||
| NM_013680.4 | NP_038708.3 | |||
| BB646141.1 | ||||
| BC022954.1 | AAH22954.1 |
More Information
Set Membership: |
|

Genomic Map