Assay Details
Target Gene Details
Entrez Gene ID: | 16784 |
Gene Name: | lysosomal-associated membrane protein 2 |
Gene Aliases: |
CD107b, LGP-B, Lamp II, Lamp-2, Lamp-2a, Lamp-2b, Lamp-2c, Mac3 |
Location: |
Chr.X:38401357-38456460 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamp2 | NM_001017959.2 | 5 | 856 | NP_001017959.1 |
| NM_001290485.1 | 5 | 856 | NP_001277414.1 | |
| NM_010685.4 | 5 | 856 | NP_034815.2 | |
| AK012149.1 | 5 | 815 | BAB28062.1 | |
| AK032974.1 | 5 | 818 | BAC28106.1 | |
| AK048174.1 | 5 | 798 | BAC33265.1 | |
| AK051989.1 | 4 | 1654 | ||
| AK078193.1 | 5 | 850 | BAC37169.1 | |
| AK088804.1 | 6 | 833 | ||
| AK128946.1 | 3 | 518 | ||
| AK136550.1 | 5 | 816 | ||
| AK146731.1 | 5 | 837 | BAE27393.1 | |
| AK150766.1 | 5 | 825 | ||
| AK159272.1 | 5 | 811 | BAE34951.1 | |
| AK159731.1 | 5 | 811 | BAE35325.1 | |
| AK163933.1 | 5 | 857 | BAE37543.1 | |
| BC138718.1 | 5 | 743 | ||
| BC138723.1 | 5 | 743 | ||
| J05287.1 | 5 | 783 | AAA39412.1 | |
| M32017.1 | 1 | 144 | AAA39429.1 |
More Information
Set Membership: |
|

Genomic Map