Assay Details
Target Gene Details
Entrez Gene ID: | 16784 |
Gene Name: | lysosomal-associated membrane protein 2 |
Gene Aliases: |
CD107b, LGP-B, Lamp II, Lamp-2, Lamp-2a, Lamp-2b, Lamp-2c, Mac3 |
Location: |
Chr.X:38401357-38456460 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamp2 | NM_001017959.2 | NP_001017959.1 | ||
| NM_001290485.1 | NP_001277414.1 | |||
| NM_010685.4 | NP_034815.2 | |||
| AK012149.1 | BAB28062.1 | |||
| AK032974.1 | BAC28106.1 | |||
| AK048174.1 | BAC33265.1 | |||
| AK078193.1 | BAC37169.1 | |||
| AK088804.1 | ||||
| AK128946.1 | ||||
| AK136550.1 | ||||
| AK146731.1 | BAE27393.1 | |||
| AK150766.1 | ||||
| AK159272.1 | BAE34951.1 | |||
| AK159731.1 | BAE35325.1 | |||
| AK163933.1 | BAE37543.1 | |||
| AK189254.1 | ||||
| BC138718.1 | ||||
| BC138723.1 | ||||
| J05287.1 | AAA39412.1 | |||
| M32017.1 | AAA39429.1 |
More Information
Set Membership: |
|

Genomic Map