Assay Details
Target Gene Details
Entrez Gene ID: | 17257 |
Gene Name: | methyl CpG binding protein 2 |
Gene Aliases: |
1500041B07Rik, D630021H01Rik, Mbd5, WBP10 |
Location: |
Chr.X:74026592-74085690 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mecp2 | NM_001081979.2 | NP_001075448.1 | ||
| NM_010788.4 | NP_034918.1 | |||
| AF158181.1 | AAF33024.1 | |||
| AJ132922.1 | CAB46495.1 | |||
| AK158664.1 | BAE34602.1 | |||
| BC027153.1 | AAH27153.1 | |||
| BY107013.1 |
More Information
Set Membership: |
|

Genomic Map