Assay Details
Target Gene Details
Entrez Gene ID: | 320923 |
Gene Name: | MAP7 domain containing 3 |
Gene Aliases: |
4933424A10Rik, Map7d3 |
Location: |
Chr.X:56775049-56822490 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtap7d3 | NM_177293.3 | NP_796267.2 | ||
| AK030211.1 | BAC26846.1 | |||
| AK048385.1 | BAC33317.1 | |||
| BC118963.2 |
More Information
Set Membership: |
|

Genomic Map