Assay Details
Target Gene Details
Entrez Gene ID: | 20977 |
Gene Name: | synaptophysin |
Gene Aliases: |
A230093K24Rik, AI848995, Syn, p38 |
Location: |
Chr.X:7638580-7653256 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syp | NM_009305.2 | 3 | 279 | NP_033331.2 |
| AK034291.1 | 3 | 222 | ||
| AK043756.1 | 3 | 176 | BAC31642.1 | |
| AK090259.1 | 3 | 176 | ||
| BC014823.1 | 3 | 149 | AAH14823.1 | |
| BC115644.1 | 2 | 132 | ||
| BC115645.1 | 2 | 131 | ||
| BY239748.1 | 3 | 279 | ||
| X95818.1 | 3 | 148 | CAA65084.1 |
More Information
Set Membership: |
|

Genomic Map