Assay Details
Target Gene Details
Entrez Gene ID: | 20977 |
Gene Name: | synaptophysin |
Gene Aliases: |
A230093K24Rik, AI848995, Syn, p38 |
Location: |
Chr.X:7638580-7653256 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syp | NM_009305.2 | 5 | 685 | NP_033331.2 |
| AK034291.1 | 5 | 628 | ||
| AK043756.1 | 5 | 582 | BAC31642.1 | |
| AK090259.1 | 5 | 583 | ||
| BC014823.1 | 5 | 555 | AAH14823.1 | |
| BC115644.1 | 4 | 538 | ||
| BC115645.1 | 4 | 537 | ||
| X95818.1 | 5 | 554 | CAA65084.1 |
More Information
Set Membership: |
|

Genomic Map