Assay Details
Target Gene Details
Entrez Gene ID: | 20832 |
Gene Name: | signal sequence receptor, delta |
Gene Aliases: |
AW743063, C78062, Trap |
Location: |
Chr.X:73787028-73790828 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssr4 | NM_001166480.1 | NP_001159952.1 | ||
| NM_009279.3 | NP_033305.1 | |||
| AK003044.1 | BAB22529.1 | |||
| AK007921.1 | BAB25352.1 | |||
| AK013723.1 | BAB28972.1 | |||
| AK142600.1 | 1 | 1728 | ||
| AK160089.1 | BAE35620.1 | |||
| BC008172.1 | AAH08172.1 | |||
| BI989805.1 | ||||
| BQ747160.1 | ||||
| CJ144362.1 | ||||
| X90582.1 | CAA62210.1 |

Genomic Map