Assay Details
Target Gene Details
Entrez Gene ID: | 67298 |
Gene Name: | G protein-coupled receptor associated sorting protein 1 |
Gene Aliases: |
2210415K24Rik, 3110031O14Rik, C87852, GASP, GASP1 |
Location: |
Chr.X:135742692-135803468 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gprasp1 | NM_001004359.2 | NP_001004359.1 | ||
| NM_001005385.1 | NP_001005385.1 | |||
| NM_026081.5 | NP_080357.4 | |||
| AK008944.1 | ||||
| AK083192.1 | BAC38803.1 | |||
| AK170837.1 | ||||
| BC085157.1 | AAH85157.1 |
More Information
Set Membership: |
|

Genomic Map