Assay Details
Target Gene Details
Entrez Gene ID: | 53332 |
Gene Name: | myotubularin related protein 1 |
Gene Aliases: |
AW049210 |
Location: |
Chr.X:71364737-71419197 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtmr1 | NM_016985.2 | 9 | 1125 | NP_058681.1 |
| AF073997.1 | 9 | 1098 | AAC77822.1 | |
| AK048714.1 | 1 | 378 | ||
| AK051390.1 | 2 | 123 | BAC34622.1 | |
| BC056376.1 | 9 | 1125 | AAH56376.1 | |
| BC057337.1 | 9 | 1119 | AAH57337.1 |
More Information
Set Membership: |
|

Genomic Map