Assay Details
Target Gene Details
Entrez Gene ID: | 67050 |
Gene Name: | NFKB activating protein |
Gene Aliases: |
2610020O08Rik, AA987160, AI849147, AL024236 |
Location: |
Chr.X:37126763-37150746 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkap | NM_025937.4 | NP_080213.3 | ||
| AK011492.1 | BAB27654.1 | |||
| AK012826.1 | BAB28497.3 | |||
| AK038344.1 | BAC29972.1 | |||
| AK076125.1 | ||||
| AK089943.1 | BAC41010.1 | |||
| AK180844.1 | ||||
| AY388959.1 | AAQ90403.1 | |||
| BC026774.1 | AAH26774.1 |

Genomic Map