Assay Details
Target Gene Details
Entrez Gene ID: | 620592 |
Gene Name: | transmembrane protein 28 |
Gene Aliases: |
EG620592, Fam155b, Tnen28 |
Location: |
Chr.X:99821069-99846345 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem28 | NM_001081283.1 | NP_001074752.1 |

Genomic Map