Assay Details
Target Gene Details
Entrez Gene ID: | 78526 |
Gene Name: | RIKEN cDNA C330013F16 gene |
Gene Aliases: |
- |
Location: |
Chr.X:139240226-139357738 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C330013F16Rik | NR_045455.1 | |||
| AK133643.1 |
Target Gene Details
Entrez Gene ID: | 22037 |
Gene Name: | tumor rejection antigen P1A |
Gene Aliases: |
Tra-P1A |
Location: |
Chr.X:139333683-139338165 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trap1a | NM_011635.1 | 3 | 1016 | NP_035765.1 |
| AK146737.1 | 3 | 831 | BAE27399.1 | |
| BC051644.1 | 3 | 809 | AAH51644.1 | |
| M36387.1 | 3 | 1016 | AAA40475.1 |
More Information
Set Membership: |
|

Genomic Map