Assay Details
Target Gene Details
Entrez Gene ID: | 209815 |
Gene Name: | TBC1 domain family, member 25 |
Gene Aliases: |
6330576A08Rik, A530047E07, DXHXS7927e, Oatl1, mMg81 |
Location: |
Chr.X:8154470-8176181 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d25 | NM_001166437.1 | NP_001159909.1 | ||
| NM_172478.3 | NP_766066.2 | |||
| AK040932.1 | BAC30751.1 | |||
| AK078145.1 | BAC37146.1 | |||
| AK166256.1 | ||||
| BC113777.1 | ||||
| BC128565.1 | ||||
| BC128566.1 |
More Information
Set Membership: |
|

Genomic Map