Assay Details
Target Gene Details
Entrez Gene ID: | 22327 |
Gene Name: | von Hippel-Lindau binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.X:75514278-75534946 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vbp1 | NM_011692.2 | NP_035822.2 | ||
| AK002282.1 | BAB21986.1 | |||
| AK008847.1 | BAB25926.1 | |||
| AK077923.1 | BAC37066.1 | |||
| AK151527.1 | BAE30475.1 | |||
| AK153069.1 | BAE31695.1 | |||
| AK159284.1 | BAE34961.1 | |||
| AK167064.1 | BAE39225.1 | |||
| AK167742.1 | BAE39780.1 | |||
| AK169185.1 | BAE40962.1 | |||
| BC132169.1 | ||||
| BC132171.1 | ||||
| BY134471.1 | ||||
| DV066998.1 | ||||
| U96760.1 | AAC23908.1 |
More Information
Set Membership: |
|

Genomic Map