Assay Details
Target Gene Details
Entrez Gene ID: | 237052 |
Gene Name: | transcription elongation factor A (SII)-like 1 |
Gene Aliases: |
0610011M09Rik, BC011290, P21, SIIR, pp21 |
Location: |
Chr.X:136707966-136709873 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tceal1 | NM_146236.1 | NP_666348.1 | ||
| BC011290.1 | AAH11290.1 | |||
| BC083157.1 | AAH83157.1 |
More Information
Set Membership: |
|

Genomic Map