Assay Details
Target Gene Details
Entrez Gene ID: | 56496 |
Gene Name: | tetraspanin 6 |
Gene Aliases: |
6720473L21Rik, AI316786, T245, TSPAN-6, Tm4sf, Tm4sf6 |
Location: |
Chr.X:133891070-133898429 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspan6 | NM_019656.3 | NP_062630.2 | ||
| AF053454.1 | AAC69711.1 | |||
| AK146673.1 | BAE27349.1 | |||
| AK149204.1 | BAE28764.1 | |||
| AK160313.1 | BAE35735.1 | |||
| AK167384.1 | BAE39476.1 | |||
| BC003733.1 | AAH03733.1 | |||
| BC110314.1 | AAI10315.1 |

Genomic Map