Assay Details
Target Gene Details
Entrez Gene ID: | 208884 |
Gene Name: | zinc finger, DHHC domain containing 9 |
Gene Aliases: |
6430508G22, 9530098M12Rik |
Location: |
Chr.X:48171967-48208702 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zdhhc9 | NM_172465.4 | NP_766053.1 | ||
| AK032233.1 | BAC27774.1 | |||
| AK032252.1 | ||||
| AK079248.1 | ||||
| BC090832.1 | AAH90832.1 |

Genomic Map