Assay Details
Target Gene Details
Entrez Gene ID: | 319535 |
Gene Name: | zinc finger protein 182 |
Gene Aliases: |
6330583I20Rik, Znf182 |
Location: |
Chr.X:21026184-21062038 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp182 | NM_001013387.2 | NP_001013405.2 | ||
| NM_001111076.1 | NP_001104546.1 | |||
| AK043973.1 | ||||
| AK045698.1 | 1 | 163 | ||
| AK155039.1 | BAE33006.1 | |||
| BC063066.1 | AAH63066.1 |
More Information
Set Membership: |
|

Genomic Map